missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human C14orf3, homolog 1 Protein
A DNA sequence encoding the Homo sapiens (Human) C14orf3, homolog 1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8507-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001308370.1 | |
C14orf3, homolog 1 Protein | |
Research Use Only | |
C14orf3 | |
Recombinant Protein | |
E. coli | |
MAKWGEGDPR WIVEERADAT NVNNWHWTER DASNWSTDKL KTLFLAVQVQ NEEGKCEVTE VSKLDGEASI NNRKGKLIFF YEWSVKLNWT GTSKSGVQYK GHVEIPNLSD ENSVDEVEIS VSLAKDEPDT NLVALMKEEG VKLLREAMGI YISTLKTEFT QGMILPTMNG ESVDPVGQPA LKTEERKAKP APSKTQARPV GVKIPTCKIT LKETFLTSPE ELYRVFTTQE LVQAFTHAPA TLEADRGGKF HMVDGNVSGE FTDLVPEKHI VMKWRFKSWP EGHFATITLT FIDKNGETEL CMEGRGIPAP EEERTRQGWQ RYYFEGIKQT FGYGARLF | |
Greater than 90% as determined by SDS-PAGE. |
10598 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |