missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human C-X-C motif chemokine 17 Protein
A DNA sequence encoding the Human C-X-C motif chemokine 17, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10220-ec-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_940879.1 | |
C-X-C motif chemokine 17 Protein | |
Research Use Only | |
CXCL17 | |
The ED50 as determined by its ability to induce VEGF expression using murine endothelial cells is less than 5.0 ug/ml, corresponding to a specific activity of > 200 IU/mg. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
284340 | |
100 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
SSLNPGVARG HRDRGQASRR WLQEGGQECE CKDWFLRAPR RKFMTVSGLP KKQCPCDHFK GNVKKTRHQR HHRKPNKHSR ACQQFLKQCQ LRSFALPL | |
Greater than 96% as determined by SDS-PAGE and HPLC. |