missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BRISC complex subunit Abro1 Protein
A DNA sequence encoding the Homo sapiens (Human) BRISC complex subunit Abro1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7572-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_115558.3 | |
BRISC complex subunit Abro1 Protein | |
Research Use Only | |
FAM175B | |
Recombinant Protein | |
E. coli | |
MAASISGYTF SAVCFHSANS NADHEGFLLG EVRQEETFSI SDSQISNTEF LQVIEIHNHQ PCSKLFSFYD YASKVNEESL DRILKDRRKK VIGWYRFRRN TQQQMSYREQ VLHKQLTRIL GVPDLVFLLF SFISTANNST HALEYVLFRP NRRYNQRISL AIPNLGNTSQ QEYKVSSVPN TSQSYAKVIK EHGTDFFDKD GVMKDIRAIY QVYNALQEKV QAVCADVEKS ERVVESCQAE VNKLRRQITQ RKNEKEQERR LQQAVLSRQM PSESLDPAFS PRMPSSGFAA EGRSTLGDAE ASDPPPPYSD FHPNNQESTL SHSRMERSVF MPRPQAVGSS NYASTSAGLK YPGSGADLPP PQRAAGDSGE DSDDSDYENL IDPTEPSNSE YSHSKDSRPM AHPDEDPRNT QTSQI | |
Greater than 90% as determined by SDS-PAGE. |
23172 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |