missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BRD1 Protein
A cDNA sequence encoding the BRD1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11202-20ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
BRD1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSEQVAMEL RLTPLTVLLR SVLDQLQDKD PARIFAQPVS LKEVPDYLDH IKHPMDFATM RKRLEAQGYK NLHEFEEDFD LIIDNCMKYN ARDTVFYRAA VRLRDQGGVV LRQARREVDS IGLEEASGMH LPERPA | |
Greater than 95.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
BRD1 protein solution (0.5 mg/ml) containing Phosphate buffered saline (pH 7.4), 10% glycerol and 1mM DTT. |