missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Bone morphogenetic protein 1
A DNA sequence encoding the Homo sapiens (Human) Bone morphogenetic protein 3, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5723-ec-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001192.3 | |
Bone morphogenetic protein 1 | |
Research Use Only | |
BMP3 | |
Recombinant Protein | |
E. coli | |
QWIEPRNCAR RYLKVDFADI GWSEWIISPK SFDAYYCSGA CQFPMPKSLK PSNHATIQSI VRAVGVVPGI PEPCCVPEKM SSLSILFFDE NKNVVLKVYP NMTVESCACR | |
Greater than 90% as determined by SDS-PAGE. |
651 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |