missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BMP-2 Protein
Recombinant Protein
Marca: enQuireBio™ QP5363-HEK-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001191.1 | |
BMP-2 Protein | |
Research Use Only | |
BMP2 | |
Recombinant Protein | |
Human (HEK293) | |
MQAKHKQRKR LKSSCKRHPL YVDFSDVGWN DWIVAPPGYH AFYCHGECPF PLADHLNSTN HAIVQTLVNS VNSKIPKACC VPTELSAISM LYLDENEKVV LKNYQDMVVE GCGCR | |
Greater than 95% as determined by SDS-PAGE. |
650 | |
10 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
E. coli synthesized BMP-2Shows bioactivity at a concentration of 250 ng/mL in the Alkaline phosphatase activity induced in ATDC-5 cells assay. The bioactivity acceptance criteria is set at 4 x 10^3 units/mg. The more potent HEK293 derived BMP2 has a specific activity of less than 40 ng/ml in ATDC-5 cell induced alk phos activity. | |
Human | |
Untagged | |
Lyophilized from a sterile filtered (0.22 um) 2x PBS solution. |