missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BEX1 Protein
A cDNA sequence encoding the BEX1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11157-10ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
55859 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMESKEKR AVNSLSMENA NQENEEKEQV ANKGEPLALP LDAGEYCVPR GNRRRFRVRQ PILQYRWDMM HRLGEPQARM REENMERIGE EVRQLMEKLR EKQLSHSLRA VSTDPPHHDH HDEFCLMP | |
Greater than 80% as determined by SDS-PAGE. |
BEX1 Protein | |
Research Use Only | |
BEX1 | |
Human | |
His | |
The BEX1 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl and 30% glycerol. |