missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Beta-crystallin S Protein
A DNA sequence encoding the Homo sapiens (Human) Beta-crystallin S, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5872-ec-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_060011.1 | |
Beta-crystallin S Protein | |
Research Use Only | |
CRYGS | |
Recombinant Protein | |
E. coli | |
SKTGTKITFY EDKNFQGRRY DCDCDCADFH TYLSRCNSIK VEGGTWAVYE RPNFAGYMYI LPQGEYPEYQ RWMGLNDRLS SCRAVHLPSG GQYKIQIFEK GDFSGQMYET TEDCPSIMEQ FHMREIHSCK VLEGVWIFYE LPNYRGRQYL LDKKEYRKPI DWGAASPAVQ SFRRIVE | |
Greater than 80% as determined by SDS-PAGE. |
1427 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |