missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Bcl 2 (minus NWGR domain) Protein
A cDNA sequence encoding the Bcl 2 (minus NWGR domain) was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10494-HIS-10ug
Additional Details : Peso : 0.01000kg
Specifications
Bcl 2 (minus NWGR domain) Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MAHAGRTGYD NREIVMKYIH YKLSQRGYEW DAGDVGAAPP GAAPAPGIFS SQPGHTPHPA ASRDPVARTS PLQTPAAPGA AAGPALSPVP PVVHLTLRQA GDDFSRRYRR DFAEMSSQLH LTPFTARGRF ATVVEELFRD GVNWGRIVAF FEFGGVMCVE SVNREMSPLV DNIALWMTEY LNRHLHTWIQ DNGGWDAFVE LYGPSMRPLF DFSWLSLKTL LSLALVGACI TLGAYLGHK | |
Greater than 95.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The protein contains 10mM Tris-HCL pH 8.0, 1mM EDTA and 250mM NaCl. |