missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Bcl 2 (minus BH2 domain) Protein
A cDNA sequence encoding the Bcl 2 (minus BH2 domain) was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10492-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
Bcl 2 (minus BH2 domain) Protein | |
100 μg | |
Untested | |
West nile virus | |
His |
>95% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
Human | |
DPBS pH 7.4. |