missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human B9D1 Protein
A cDNA sequence encoding the B9D1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11120-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
B9D1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMATASPS VFLLMVNGQV ESAQFPEYDD LYCKYCFVYG QDWAPTAGLE EGISQITSKS QDVRQALVWN FPIDVTFKST NPYGWPQIVL SVYGPDVFGN DVVRGYGAVH VPFSPGRHKR TIPMFVPEST SKLQKFTSWF MGRRPEYTDP KVVAQGEGRE VTRVRSQGFV TLLFNVVTKD MRKLGYDTGP SDTQGVLGPS PPQSFPQ | |
Greater than 85% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
B9D1 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.4M Urea. |