Learn More
enQuireBio™ Recombinant Human Atrial natriuretic peptide receptor 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Atrial natriuretic peptide receptor 1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6431-ye-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_000897.3 | |
Atrial natriuretic peptide receptor 1 Protein | |
500 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
4881 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
NPR1 | |
Recombinant Protein | |
Yeast | |
GNLTVAVVLP LANTSYPWSW ARVGPAVELA LAQVKARPDL LPGWTVRTVL GSSENALGVC SDTAAPLAAV DLKWEHNPAV FLGPGCVYAA APVGRFTAHW RVPLLTAGAP ALGFGVKDEY ALTTRAGPSY AKLGDFVAAL HRRLGWERQA LMLYAYRPGD EEHCFFLVEG LFMRVRDRLN ITVDHLEFAE DDLSHYTRLL RTMPRKGRVI YICSSPDAFR TLMLLALEAG LCGEDYVFFH LDIFGQSLQG GQGPAPRRPW ERGDGQDVSA RQAFQAAKII TYKDPDNPEY LEFLKQLKHL AYEQFNFTME DGLVNTIPAS FHDGLLLYIQ AVTETLAHGG TVTDGENITQ RMWNRSFQGV TGYLKIDSSG DRETDFSLWD MDPENGAFRV VLNYNGTSQE LVAVSGRKLN WPLGYPPPDI PKCGFDNEDP ACNQDHLSTL E |