missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ATPase ASNA1 Protein
A DNA sequence encoding the Homo sapiens (Human) ATPase ASNA1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5685-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_004308.2 | |
ATPase ASNA1 Protein | |
Research Use Only | |
ASNA1 | |
Recombinant Protein | |
E. coli | |
AAGVAGWGVE AEEFEDAPDV EPLEPTLSNI IEQRSLKWIF VGGKGGVGKT TCSCSLAVQL SKGRESVLII STDPAHNISD AFDQKFSKVP TKVKGYDNLF AMEIDPSLGV AELPDEFFEE DNMLSMGKKM MQEAMSAFPG IDEAMSYAEV MRLVKGMNFS VVVFDTAPTG HTLRLLNFPT IVERGLGRLM QIKNQISPFI SQMCNMLGLG DMNADQLASK LEETLPVIRS VSEQFKDPEQ TTFICVCIAE FLSLYETERL IQELAKCKID THNIIVNQLV FPDPEKPCKM CEARHKIQAK YLDQMEDLYE DFHIVKLPLL PHEVRGADKV NTFSALLLEP YKPPSAQ | |
Greater than 90% as determined by SDS-PAGE. |
439 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |