missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ATP2A2 Protein
A DNA sequence encoding the Homo sapiens (Human) ATP2A2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5691-ec-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001672.1 | |
ATP2A2 Protein | |
Research Use Only | |
ATP2A2 | |
Recombinant Protein | |
E. coli | |
VITTCLALGT RRMAKKNAIV RSLPSVETLG CTSVICSDKT GTLTTNQMSV CRMFILDRVE GDTCSLNEFT ITGSTYAPIG EVHKDDKPVN CHQYDGLVEL ATICALCNDS ALDYNEAKGV YEKVGEATET ALTCLVEKMN VFDTELKGLS KIERANACNS VIKQLMKKEF TLEFSRDRKS MSVYCTPNKP SRTSMSKMFV KGAPEGVIDR CTHIRVGSTK VPMTSGVKQK IMSVIREWGS GSDTLRCLAL ATHDNPLRRE EMHLEDSANF IKYETNLTFV GCVGMLDPPR IEVASSVKLC RQAGIRVIMI TGDNKGTAVA ICRRIGIFGQ DEDVTSKAFT GREFDELNPS AQRDACLNAR CFARVEPSHK SKIVEFLQSF DEITAMTGDG VNDAPALKKA EIGIAMGSGT AVAKTASEMV LADDNFSTIV AAVEEGRAIY NNM | |
Greater than 90% as determined by SDS-PAGE. |
488 | |
100 μg | |
Not determined. | |
Cytoplasmic Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |