missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ATP synthase subunit d, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) ATP synthase subunit d, mitochondrial, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5696-ec-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001003785.1 | |
ATP synthase subunit d, mitochondrial Protein | |
Research Use Only | |
ATP5H | |
Recombinant Protein | |
E. coli | |
AGRKLALKTI DWVAFAEIIP QNQKAIASSL KSWNETLTSR LAALPENPPA IDWAYYKANV AKAGLVDDFE KKFNALKVPV PEDKYTAQVD AEEKEDVKSC AEWVSLSKAR IVEYEKEMEK MKNLIPFDQM TIEDLNEAFP ETKLDKKKYP YWPHQPIENL | |
Greater than 80% as determined by SDS-PAGE. |
10476 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |