missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ATP-binding cassette sub-family D1 Protein
A DNA sequence encoding the Homo sapiens (Human) ATP-binding cassette sub-family D member 1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP9947-iv-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_000024.2 | |
ATP-binding cassette sub-family D1 Protein | |
Research Use Only | |
ABCD1 | |
Recombinant Protein | |
E. coli | |
MPVLSRPRPW RGNTLKRTAV LLALAAYGAH KVYPLVRQCL APARGLQAPA GEPTQEASGV AAAKAGMNRV FLQRLLWLLR LLFPRVLCRE TGLLALHSAA LVSRTFLSVY VARLDGRLAR CIVRKDPRAF GWQLLQWLLI ALPATFVNSA IRYLEGQLAL SFRSRLVAHA YRLYFSQQTY YRVSNMDGRL RNPDQSLTED VVAFAASVAH LYSNLTKPLL DVAVTSYTLL RAARSRGAGT AWPSAIAGLV VFLTANVLRA FSPKFGELVA EEARRKGELR YMHSRVVANS EEIAFYGGHE VELALLQRSY QDLASQINLI LLERLWYVML EQFLMKYVWS ASGLLMVAVP IITATGYSES DAEAVKKAAL EKKEEELVSE RTEAFTIARN LLTAAADAIE RIMSSYKEVT ELAGYTARVH EMFQVFEDVQ RCHFKRPREL EDAQAGSGTI GRSGVRVEGP LKIRGQVVDV EQGIICENIP IVTPSGEVVV ASLNIRVEEG MHLLITGPNG CGKSSLFRIL GGLWPTYGGV LYKPPPQRMF YIPQRPYMSV GSLRDQVIYP DSVEDMQRKG YSEQDLEAIL DVVHLHHILQ REGGWEAMCD WKDVLSGGEK QRIGMARMFY HRPKYALLDE CTSAVSIDVE GKIFQAAKDA GIALLSITHR PSLWKYHTHL LQFDGEGGWK FEKLDSAARL SLTEEKQRLE QQLAGIPKMQ RRLQELCQIL GEAVAPAHVP APSPQGPGGL QGASTDDDDK HHHHHHHHHH | |
Greater than 85% as determined by SDS-PAGE. |
215 | |
100 μg | |
Not determined. | |
Full Length Transmembrane Protein | |
Human | |
His | |
Tris-based buffer, 50% glycerol |