missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Aquaporin-1 Protein
A DNA sequence encoding the Homo sapiens (Human) Aquaporin-1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5671-ec-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_932766.1 | |
Aquaporin-1 Protein | |
Research Use Only | |
AQP1 | |
Recombinant Protein | |
E. coli | |
GALAVLIYDF ILAPRSSDLT DRVKVWTSGQ VEEYDLDADD INSRVEMKPK | |
Greater than 90% as determined by SDS-PAGE. |
358 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |