missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human APOM Protein
A DNA sequence encoding the Homo sapiens (Human) APOM, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5669-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001243098.1 | |
APOM Protein | |
Research Use Only | |
APOM | |
Recombinant Protein | |
E. coli | |
MFHQIWAALL YFYGIILNSI YQCPEHSQLT TLGVDGKEFP EVHLGQWYFI AGAAPTKEEL ATFDPVDNIV FNMAAGSAPM QLHLRATIRM KDGLCVPRKW IYHLTEGSTD LRTEGRPDMK TELFSSSCPG GIMLNETGQG YQRFLLYNRS PHPPEKCVEE FKSLTSCLDS KAFLLTPRNQ EACELSNN | |
Greater than 90% as determined by SDS-PAGE. |
55937 | |
10 μg | |
Not determined. | |
Full Length of Mature Protein | |
Human | |
His-GST | |
Tris-based buffer, 50% glycerol |