missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Apolipoprotein C-III Protein
A DNA sequence encoding the Homo sapiens (Human) Apolipoprotein C-III, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5666-ec-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_000031.1 | |
Apolipoprotein C-III Protein | |
Research Use Only | |
APOC3 | |
Recombinant Protein | |
E. coli | |
SEAEDASLLS FMQGYMKHAT KTAKDALSSV QESQVAQQAR GWVTDGFSSL KDYWSTVKDK FSEFWDLDPE VRPTSAVAA | |
Greater than 90% as determined by SDS-PAGE. |
345 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |