missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Apolipoprotein C-II Protein
A DNA sequence encoding the Homo sapiens (Human) Apolipoprotein C-II, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5665-ye-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_000474.2 | |
Apolipoprotein C-II Protein | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
344 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
APOC2 | |
Recombinant Protein | |
Yeast | |
TQQPQQDEMP SPTFLTQVKE SLSSYWESAK TAAQNLYEKT YLPAVDEKLR DLYSKSTAAM STYTGIFTDQ VLSVLKGEE |