missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Antigen-presenting glycoprotein CD1d Protein
A DNA sequence encoding the Homo sapiens (Human) Antigen-presenting glycoprotein CD1d, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5784-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001306074.1 | |
Antigen-presenting glycoprotein CD1d Protein | |
Research Use Only | |
CD1D | |
Recombinant Protein | |
E. coli | |
EVPQRLFPLR CLQISSFANS SWTRTDGLAW LGELQTHSWS NDSDTVRSLK PWSQGTFSDQ QWETLQHIFR VYRSSFTRDV KEFAKMLRLS YPLELQVSAG CEVHPGNASN NFFHVAFQGK DILSFQGTSW EPTQEAPLWV NLAIQVLNQD KWTRETVQWL LNGTCPQFVS GLLESGKSEL KKQVKPKAWL SRGPSPGPGR LLLVCHVSGF YPKPVWVKWM RGEQEQQGTQ PGDILPNADE TWYLRATLDV VAGEAAGLSC RVKHSSLEGQ DIVLYWGGSY TS | |
Greater than 90% as determined by SDS-PAGE. |
912 | |
10 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |