missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Amelogenin, X isoform Protein
A DNA sequence encoding the Homo sapiens (Human) Amelogenin, X isoform, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8035-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001133.1 | |
Amelogenin, X isoform Protein | |
Research Use Only | |
AMELX | |
Recombinant Protein | |
E. coli | |
MPLPPHPGHP GYINFSYEVL TPLKWYQSIR PPYPSYGYEP MGGWLHHQII PVLSQQHPPT HTLQPHHHIP VVPAQQPVIP QQPMMPVPGQ HSMTPIQHHQ PNLPPPAQQP YQPQPVQPQP HQPMQPQPPV HPMQPLPPQP PLPPMFPMQP LPPMLPDLTL EAWPSTDKTK REEVD | |
Greater than 90% as determined by SDS-PAGE. |
265 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |