missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ALDOB Protein
A cDNA sequence encoding the ALDOB was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11002-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
229 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMAHRFP ALTQEQKKEL SEIAQSIVAN GKGILAADES VGTMGNRLQR IKVENTEENR RQFREILFSV DSSINQSIGG VILFHETLYQ KDSQGKLFRN ILKEKGIVVG IKLDQGGAPL AGTNKETTIQ GLDGLSERCA QYKKDGVDFG KWRAVLRIAD QCPSSLAIQE NANALARYAS ICQQNGLVPI VEPEVIPDGD HDLEHCQYVT EKVLAAVYKA LNDHHVYLEG TLLKPNMVTA GHACTKKYTP EQVAMATVTA LHRTVPAAVP GICFLSGGMS EEDATLNLNA INLCPLPKPW KLSFSYGRAL QASALAAWGG KAANKEATQE AFMKRAMANC QAAKGQYVHT GSSGAASTQS LFTACYTY | |
Greater than 95.0% as determined by SDS-PAGE. |
ALDOB Protein | |
Research Use Only | |
ALDOB | |
Human | |
His | |
The ALDOB solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 10% glycerol and 0.1M NaCl. |