missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ALDH1A3 / RALDH3 Protein
A DNA sequence encoding the Homo sapiens (Human) ALDH1A3 / RALDH3, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5644-ye-500ug
Additional Details : Peso : 0.01000kg
Specifications
NP_000684.2 | |
ALDH1A3 / RALDH3 Protein | |
Research Use Only | |
ALDH1A3 | |
Recombinant Protein | |
Yeast | |
ATANGAVENG QPDRKPPALP RPIRNLEVKF TKIFINNEWH ESKSGKKFAT CNPSTREQIC EVEEGDKPDV DKAVEAAQVA FQRGSPWRRL DALSRGRLLH QLADLVERDR ATLAALETMD TGKPFLHAFF IDLEGCIRTL RYFAGWADKI QGKTIPTDDN VVCFTRHEPI GVCGAITPWN FPLLMLVWKL APALCCGNTM VLKPAEQTPL TALYLGSLIK EAGFPPGVVN IVPGFGPTVG AAISSHPQIN KIAFTGSTEV GKLVKEAASR SNLKRVTLEL GGKNPCIVCA DADLDLAVEC AHQGVFFNQG QCCTAASRVF VEEQVYSEFV RRSVEYAKKR PVGDPFDVKT EQGPQIDQKQ FDKILELIES GKKEGAKLEC GGSAMEDKGL FIKPTVFSEV TDNMRIAKEE IFGPVQPILK FKSIEEVIKR ANSTDYGLTA AVFTKNLDKA LKLASALESG TVWINCYNAL YAQAPFGGFK MSGNGRELGE YALAEYTEVK TVTIKLGDKN P | |
Greater than 90% as determined by SDS-PAGE. |
220 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |