missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Alcohol dehydrogenase 4 Protein
A DNA sequence encoding the Homo sapiens (Human) Alcohol dehydrogenase 4, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5615-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_000661.2 | |
Alcohol dehydrogenase 4 Protein | |
Research Use Only | |
ADH4 | |
Recombinant Protein | |
E. coli | |
MGTKGKVIKC KAAIAWEAGK PLCIEEVEVA PPKAHEVRIQ IIATSLCHTD ATVIDSKFEG LAFPVIVGHE AAGIVESIGP GVTNVKPGDK VIPLYAPLCR KCKFCLSPLT NLCGKISNLK SPASDQQLME DKTSRFTCKG KPVYHFFGTS TFSQYTVVSD INLAKIDDDA NLERVCLLGC GFSTGYGAAI NNAKVTPGST CAVFGLGGVG LSAVMGCKAA GASRIIGIDI NSEKFVKAKA LGATDCLNPR DLHKPIQEVI IELTKGGVDF ALDCAGGSET MKAALDCTTA GWGSCTFIGV AAGSKGLTIF PEELIIGRTI NGTFFGGWKS VDSIPKLVTD YKNKKFNLDA LVTHTLPFDK ISEAFDLMNQ GKSVRTILIF | |
Greater than 80% as determined by SDS-PAGE. |
127 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |