missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human AKR1C2 Protein
A DNA sequence encoding the Homo sapiens (Human) AKR1C2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5637-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001128713.1 | |
AKR1C2 Protein | |
Research Use Only | |
AKR1C2 | |
Recombinant Protein | |
E. coli | |
MDSKYQCVKL NDGHFMPVLG FGTYAPAEVP KSKALEAVKL AIEAGFHHID SAHVYNNEEQ VGLAIRSKIA DGSVKREDIF YTSKLWSNSH RPELVRPALE RSLKNLQLDY VDLYLIHFPV SVKPGEEVIP KDENGKILFD TVDLCATWEA MEKCKDAGLA KSIGVSNFNH RLLEMILNKP GLKYKPVCNQ VECHPYFNQR KLLDFCKSKD IVLVAYSALG SHREEPWVDP NSPVLLEDPV LCALAKKHKR TPALIALRYQ LQRGVVVLAK SYNEQRIRQN VQVFEFQLTS EEMKAIDGLN RNVRYLTLDI FAGPPNYPFS DEY | |
Greater than 90% as determined by SDS-PAGE. |
1646 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |