missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human AKR1C1 Protein

A cDNA sequence encoding the AKR1C1 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP10513-5ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15994098

Cantidad Precio
1 129.74€ / 5 microgramos
10 121.33€ / 5 microgramos
Envío estimado: 01-03-2022

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.



Descripción

Descripción

Especificaciones

Especificaciones

AKR1C1 Protein
5 μg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MDSKYQCVKL NDGHFMPVLG FGTYAPAEVP KSKALEATKL AIEAGFRHID SAHLYNNEEQ VGLAIRSKIA DGSVKREDIF YTSKLWCNSH RPELVRPALE RSLKNLQLDY VDLYLIHFPV SVKPGEEVIP KDENGKILFD TVDLCATWEA VEKCKDAGLA KSIGVSNFNR RQLEMILNKP GLKYKPVCNQ VECHPYFNQR KLLDFCKSKD IVLVAYSALG SHREEPWVDP NSPVLLEDPV LCALAKKHKR TPALIALRYQ LQRGVVVLAK SYNEQRIRQN VQVFEFQLTS EEMKAIDGLN RNVRYLTLDI FAGPPNYPFS DEY
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Specific activity is > 500 pmol/min/ug, and is defined as the amount of enzyme that catalyze the oxidation of 1.0 pmole 1-Acenaphthenol in the presence of NADP per minute at pH 8.8 at 25C.
Human
His
The AKR1C1 protein solution (0.5 mg/ml) contains 20mM Tris-HCl pH 8.0, 1mM DTT and 20% glycerol.
Videos
SDS
Documentos
Certificados
Ofertas especiales

Ofertas especiales