missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human aFGF / FGF1 Protein
A DNA sequence encoding the Homo sapiens (Human) aFGF / FGF1 Protein, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6036-ec-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_000791.1 | |
aFGF / FGF1 Protein | |
Research Use Only | |
FGF1 | |
Recombinant Protein | |
E. coli | |
FNLPPGNYKK PKLLYCSNGG HFLRILPDGT VDGTRDRSDQ HIQLQLSAES VGEVYIKSTE TGQYLAMDTD GLLYGSQTPN EECLFLERLE ENHYNTYISK KHAEKNWFVG LKKNGSCKRG PRTHYGQKAI LFLPLPVSSD | |
Greater than 90% as determined by SDS-PAGE. |
2246 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |