missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ACYP1 Protein
A cDNA sequence encoding the ACYP1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10959-20ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
ACYP1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAEGNTL ISVDYEIFGK VQGVFFRKHT QAEGKKLGLV GWVQNTDRGT VQGQLQGPIS KVRHMQEWLE TRGSPKSHID KANFNNEKVI LKLDYSDFQI VK | |
Greater than 90.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The ACYP1 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 20% glycerol, 0.1M NaCl and 1mM DTT. |