missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Activin A / INHBA Protein
Recombinant Protein
Marca: enQuireBio™ QP5357-2ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_002183.1 | |
Activin A / INHBA Protein | |
Research Use Only | |
INHBA | |
Recombinant Protein | |
E. coli | |
MGLECDGKVN ICCKKQFFVS FKDIGWNDWI IAPSGYHANY CEGECPSHIA GTSGSSLSFH STVINHYRMR GHSPFANLKS CCVPTKLRPM SMLYYDDGQN IIKKDIQNMI VEECGCS | |
Greater than 95% as determined by SDS-PAGE. |
3624 | |
2 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Shows bioactivity at a concentration of 10 ng/mL in the Cytotoxicity of MPC-11 cells assay. The bioactivity acceptance criteria is set at 1 x 10^5 units/mg. | |
Human | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |