missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Actin, cytoplasmic 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Actin, cytoplasmic 1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8379-ec-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001092.1 | |
Actin, cytoplasmic 1 Protein | |
Research Use Only | |
ACTB | |
Recombinant Protein | |
E. coli | |
PVLLTEAPLN PKANREKMTQ IMFETFNTPA MYVAIQAVLS LYASGRTTGI VMDSGDGVTH TVPIYEGYAL PHAILRLDLA GRDLTDYLMK ILTERGYSFT TTAEREIVRD IKEKLCYVAL DFEQEMATAA SSSSLEKSYE LPDGQVITIG NERFRCPEAL FQPSFLGMES CGIHETTFNS IMKCDVDIRK DLYANTVLSG GTTMYPGIAD RMQKEITALA PSTMKIKIIA PPERKYSVWI GGSILASLST FQQMWISKQE YDESGPSIVH RKCF | |
Greater than 90% as determined by SDS-PAGE. |
60 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |