missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ACLY / acly / ATP citrate lyase Protein
A DNA sequence encoding the Homo sapiens (Human) ACLY / acly / ATP citrate lyase, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5607-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Specifications
NP_001087.2 | |
ACLY / acly / ATP citrate lyase Protein | |
Research Use Only | |
ACLY | |
Recombinant Protein | |
E. coli | |
KAISEQTGKE LLYKFICTTS AIQNRFKYAR VTPDTDWARL LQDHPWLLSQ NLVVKPDQLI KRRGKLGLVG VNLTLDGVKS WLKPRLGQEA TVGKATGFLK NFLIEPFVPH SQAEEFYVCI YATREGDYVL FHHEGGVDVG DVDAKAQKLL VGVDEKLNPE DIKKHLLVHA PEDKKEILAS FISGLFNFYE DLYFTYLEIN PLVVTKDGVY VLDLAAKVDA TADYICKVKW GDIEFPPPFG REAYPEEAYI ADLDAKSGAS LK | |
Greater than 90% as determined by SDS-PAGE. |
47 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |