missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ACADS Protein
A cDNA sequence encoding the ACADS was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10933-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
35 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MLHTIYQSVE LPETHQMLLQ TCRDFAEKEL FPIAAQVDKE HLFPAAQVKK MGGLGLLAMD VPEELGGAGL DYLAYAIAME EISRGCASTG VIMSVNNSLY LGPILKFGSK EQKQAWVTPF TSGDKIGCFA LSEPGNGSDA GAASTTARAE GDSWVLNGTK AWITNAWEAS AAVVFASTDR ALQNKSISAF LVPMPTPGLT LGKKEDKLGI RGSSTANLIF EDCRIPKDSI LGEPGMGFKI AMQTLDMGRI GIASQALGIA QTALDCAVNY AENRMAFGAP LTKLQVIQFK LADMALALES ARLLTWRAAM LKDNKKPFIK EAAMAKLAAS EAATAISHQA IQILGGMGYV TEMPAERHYR DARITEIYEG TSEIQRLVIA GHLLRSYRS | |
Greater than 95.0% as determined by SDS-PAGE. |
ACADS Protein | |
Research Use Only | |
ACADS | |
Human | |
His | |
The protein contains 20mM Tris buffer pH 8.0, 1mM DTT, 20% glycerol and 100mM NaCl. |