missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human 60S acidic ribosomal protein P1 Protein
A DNA sequence encoding the Homo sapiens (Human) 60S acidic ribosomal protein P1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6625-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_000994.1 | |
60S acidic ribosomal protein P1 Protein | |
Research Use Only | |
RPLP1 | |
Recombinant Protein | |
E. coli | |
ASVSELACIY SALILHDDEV TVTEDKINAL IKAAGVNVEP FWPGLFAKAL ANVNIGSLIC NVGAGGPAPA AGAAPAGGPA PSTAAAPAEE KKVEAKKEES EESDDDMGFG LFD | |
Greater than 90% as determined by SDS-PAGE. |
6176 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |