missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human 3-ketoacyl-CoA thiolase, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) 3-ketoacyl-CoA thiolase, mitochondrial, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5603-ye-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_006102.2 | |
3-ketoacyl-CoA thiolase, mitochondrial Protein | |
Research Use Only | |
ACAA2 | |
Recombinant Protein | |
Yeast | |
FGAYGGLLKD FTATDLSEFA AKAALSAGKV SPETVDSVIM GNVLQSSSDA IYLARHVGLR VGIPKETPAL TINRLCGSGF QSIVNGCQEI CVKEAEVVLC GGTESMSQAP YCVRNVRFGT KLGSDIKLED SLWVSLTDQH VQLPMAMTAE NLAVKHKISR EECDKYALQS QQRWKAANDA GYFNDEMAPI EVKTKKGKQT MQVDEHARPQ TTLEQLQKLP PVFKKDGTVT AGNASGVADG AGAVIIASED AVKKHNFTPL ARIVGYFVSG CDPSIMGIGP VPAISGALKK AGLSLKDMDL VEVNEAFAPQ YLAVERSLDL DISKTNVNGG AIALGHPLGG SGSRITAHLV HELRRRGGKY AVGSACIGGG QGIAVIIQST A | |
Greater than 90% as determined by SDS-PAGE. |
10449 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |