missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant HPV HPV 16 Protein
A cDNA sequence encoding the HPV 16 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12307-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
HPV HPV 16 Protein | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
HPV | |
GST | |
Recombinant HPV-16 solution in PBS, 100mM arginine and 3M Urea. |
Protein is >90% pure as determined by 10% SDS-PAGE (Coomassie staining). | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MQVTFIYILVITCYENDVNVYHIFFQMSLWLPSEATVYLPPVPVSKVVSTDEYVARTNIYYHAGTSRLLAVGHPYFPIKKPNNNKILVPKVSGLQYRVFRIHLPDPNKFGFPDTSFYNPDTQRLVWACVGVEVGRGQPLGVGISGHPLLNKLDDTENASAYAANAGVDNRECISMDYKQTQLCLIGCKPPIGEHWGKGSPCTNVAVNPGDCPPLELINTVIQDGDMVHTGFGAMDFTTLQANKSEVPLDICTSICKYPDYIKMVSEPYGDSLFFYLRREQMFVRHLFNRAGTVGENVPDDLYIKGSGSTANLASSNYFPTPSGSMVTSDAQIFNKPYWLQRAQGHNNGICWGNQLFVTVVDTTRSTNMSLCAAISTSETTYKNTNFKEYLRHGEEYDLQFIFQLCKITLTADVMTYIHSMNSTILEDWNFGLQPPPGGTLEDTYRFVTQAIACQKHTPPAPKEDDPLKKYTFWEVNLKEKFSADLDQFPLGRKFLLQAGLKAKPKFTLGKRKATPTTSSTSTTAKRKKRKL |