missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Horse IL-1 beta / IL1B Protein
A DNA sequence encoding the Horse IL-1 beta / IL1B, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10471-ec-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001075995.1 | |
IL-1 beta / IL1B Protein | |
Research Use Only | |
IL1B | |
The ED50 as determined by a cell proliferation assay using murine D10S cells is less than 20 pg/ml, corresponding to a specific activity of > 5.0 M 107 IU/mg. | |
Horse | |
Untagged | |
0.2 um filtered PBS, pH 7.4, 0.1 % Tween-80, lyophilized |
100034237 | |
100 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
AAMHSVNCRL RDIYHKSLVL SGACELQAVH LNGENTNQQV VFCMSFVQGE EETDKIPVAL GLKEKNLYLS CGMKDGKPTL QLETVDPNTY PKRKMEKRFV FNKMEIKGNV EFESAMYPNW YISTSQAEKS PVFLGNTRGG RDITDFIMEI TSA | |
Greater than 95% as determined by SDS-PAGE and HPLC. |