missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Hepatitis C HCV NS5 Genotype-2 Protein
A cDNA sequence encoding the HCV NS5 Genotype-2 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12189-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
Hepatitis C HCV NS5 Genotype-2 Protein | |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Hepatitis C | |
Untagged |
HCV NS5 Genotype-2 protein is >95% pure as determined by 10% PAGE (coomassie staining). | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
25mM Tris and 1mM EDTA, 1.5M urea and 50% glycerol. |