missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Hepatitis C HCV NS3 Genotype-5a Protein
A cDNA sequence encoding the HCV NS3 Genotype-5a was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12172-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
Hepatitis C HCV NS3 Genotype-5a Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
25mM Tris-HCl, 1mM EDTA, 1.5M urea and 50% glycerol. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Hepatitis C | |
GST | |
HCV NS3 Genotype-5a protein is >95% pure as determined by 10% PAGE (coomassie staining). |