Learn More
enQuireBio™ Recombinant Hepatitis C HCV Core Genotype-3b Protein
A cDNA sequence encoding the HCV Core Genotype-3b was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12152-100ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
Hepatitis C HCV Core Genotype-3b Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
25mM Tris-Hcl pH 8.0, 0.2% Triton-X, 1.5M urea and 50% glycerol. |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Hepatitis C | |
GST | |
HCV Core Genotype-3b protein is >95% pure as determined by 10% PAGE (coomassie staining). |