missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Hepatitis C HCV Core Genotype-2b Protein
A cDNA sequence encoding the HCV Core Genotype-2b was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12149-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
Hepatitis C HCV Core Genotype-2b Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
25mM Tris-HCl, pH 8.0, 1.5M Urea, 0.2% Triton-X and 50% glycerol. |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Hepatitis C | |
GST | |
HCV Core Genotype-2b protein is >95% pure as determined by 10% PAGE (coomassie staining). |