missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Epstein-Barr Latent membrane protein 1
A DNA sequence encoding the Epstein-Barr virus (strain B95-8) (HHV-4) (Human herpesvirus 4) Latent membrane protein 2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7054-ye-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
Epstein-Barr Latent membrane protein 1 | |
Research Use Only | |
Full Length | |
Rotavirus | |
His | |
Tris-based buffer, 50% glycerol |
10 μg | |
Not determined. | |
Recombinant Protein | |
Yeast | |
AQVSEDIIIS KTSLWKEMQY NRDIIIRFKF NNSIIKLGGL GYKWSEISFK AANYQYNYLR DGEQVTAHTT CSVNGVNNFS YNGGLLPTHF SISRYEVIKE NSYVYVDYWD DSQAFRNMVY VRSLAANLNS VKCSGGNYNF QMPVGAWPVM SGGAVSLHFA GVTLSTQFTD FVSLNSLRFR FSLTVEEPPF SILRTRVSGL YGLPASNPNS GHEYYEIAGR FSLISLVPSN DDY | |
Greater than 90% as determined by SDS-PAGE. |