Learn More
enQuireBio™ Recombinant Epstein-Barr Envelope glycoprotein GP350 Protein
A DNA sequence encoding the Epstein-Barr virus (strain B95-8) (HHV-4) (Human herpesvirus 4) Envelope glycoprotein GP350, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7351-ec-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
YP_401667.1 | |
Envelope glycoprotein GP350 Protein | |
100 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Epstein-Barr | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
3783713 | |
Greater than 80% as determined by SDS-PAGE. | |
Research Use Only | |
BLLF1 | |
Recombinant Protein | |
E. coli | |
MEAALLVCQY TIQSLIHLTG EDPGFFNVEI PEFPFYPTCN VCTADVNVTI NFDVGGKKHQ LDLDFGQLTP HTKAVYQPRG AFGGSENATN LFLLELLGAG ELALTMRSKK LPINVTTGEE QQVSLESVDV YFQDVFGTMW CHHAEMQNPV YLIPETVPYI KWDNCNSTNI TAVVRAQGLD VTLPLSLPTS AQDSNFSVKT EMLGNEIDIE CIMEDGEISQ VLPGDNKFNI TCSGYESHVP SGGILTSTSP VATPIPGTGY AYSLRLTPRP VSRFLGNNSI LYVFYSGNGP KASGGDYCIQ SNIVFSDEIP ASQDMPTNTT DITYVGDNAT YSVPMVTSED ANSPNVTVTA FWAWPNNTET DFKCKWTLTS GTPSGCENIS GAFASNRTFD ITVSGLGTAP KTLIITRTAT NATTTTHKVI FSKAPESTTT SPTLNTTGFA DPNTTTGLPS STHVPTNLTA PASTGPTVST ADVTSPTPAG TTSGASPVTP SPS |