missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Transcriptional regulatory protein PhoP Protein
A DNA sequence encoding the Escherichia coli (strain K12) Transcriptional regulatory protein PhoP, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP9558-ye-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_415648.1 | |
Transcriptional regulatory protein PhoP Protein | |
Research Use Only | |
phoP | |
Recombinant Protein | |
Yeast | |
MRVLVVEDNA LLRHHLKVQI QDAGHQVDDA EDAKEADYYL NEHIPDIAIV DLGLPDEDGL SLIRRWRSND VSLPILVLTA RESWQDKVEV LSAGADDYVT KPFHIEEVMA RMQALMRRNS GLASQVISLP PFQVDLSRRE LSINDEVIKL TAFEYTIMET LIRNNGKVVS KDSLMLQLYP DAELRESHTI DVLMGRLRKK IQAQYPQEVI TTVRGQGYLF ELR | |
Greater than 80% as determined by SDS-PAGE. |
945697 | |
1 mg | |
Not determined. | |
Full. Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |