missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli 50S ribosomal protein L9 Protein
A DNA sequence encoding the Escherichia coli (strain K12) 50S ribosomal protein L9, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8623-ec-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_418624.1 | |
50S ribosomal protein L9 Protein | |
Research Use Only | |
rplI | |
Recombinant Protein | |
E. coli | |
MQVILLDKVA NLGSLGDQVN VKAGYARNFL VPQGKAVPAT KKNIEFFEAR RAELEAKLAE VLAAANARAE KINALETVTI ASKAGDEGKL FGSIGTRDIA DAVTAAGVEV AKSEVRLPNG VLRTTGEHEV SFQVHSEVFA KVIV | |
Greater than 90% as determined by SDS-PAGE. |
948720 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
E. coli | |
GST | |
Tris-based buffer, 50% glycerol |