missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli 50S ribosomal protein L35 Protein
A DNA sequence encoding the Escherichia coli (strain K12) 50S ribosomal protein L35, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8626-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_416232.3 | |
50S ribosomal protein L35 Protein | |
Research Use Only | |
rpmI | |
Recombinant Protein | |
E. coli | |
VRGAAKRFKK TGKGGFKHKH ANLRHILTKK ATKRKRHLRP KAMVSKGDLG LVIACLPY | |
Greater than 90% as determined by SDS-PAGE. |
32367223 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
E. coli | |
GST | |
Tris-based buffer, 50% glycerol |