missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli 50S ribosomal protein L30 Protein
A DNA sequence encoding the Escherichia coli (strain K12) 50S ribosomal protein L30, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8633-ec-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_417761.1 | |
50S ribosomal protein L30 Protein | |
Research Use Only | |
rpmD | |
Recombinant Protein | |
E. coli | |
MAKTIKITQT RSAIGRLPKH KATLLGLGLR RIGHTVERED TPAIRGMINA VSFMVKVEE | |
Greater than 90% as determined by SDS-PAGE. |
947797 | |
100 μg | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |