missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Dog BCHE / Butyrylcholinesterase Protein
A DNA sequence encoding the Canis lupus familiaris (Dog) (Canis familiaris) BCHE / Butyrylcholinesterase, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5708-ye-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
BCHE / Butyrylcholinesterase Protein | |
Research Use Only | |
BCHE | |
Recombinant Protein | |
Yeast | |
NTDQSFPGFP GSEMWNPNTD LSEDCLYLNV WIPTPKPKNA TVMIWIYGGG FQTGTSSLPV YDGKFLARVE RVIVVSVNYR VGALGFLALP GNPEAPGNLG LFDQQLALQW VQKNIAAFGG NPKSVTLFGE SAGAGSVGLH L | |
Greater than 90% as determined by SDS-PAGE. |
500 μg | |
Not determined. | |
Full Length | |
Dog | |
His | |
Tris-based buffer, 50% glycerol |