missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Dog Annexin A13 Protein
A DNA sequence encoding the Canis lupus familiaris (Dog) Annexin A13, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5651-ec-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001003255.1 | |
Annexin A13 Protein | |
Research Use Only | |
ANXA13 | |
Recombinant Protein | |
E. coli | |
MGNRHAKAKS HHGFDVDHDA KKLNKACKGM GTDEAAIIEI LSSRTSDERQ QIKQKYKATY GKDLEEVFKS DLSGNFEKTA LALLDRPSEY DARQLQKAMK GLGTDEAVLI EILCTRTNKE IMAIKEAYQR LFDRSLESDV KADTSGNLKA ILVSLLQANR DEGDDVDKDL AGQDAKDLYD AGDGRWGTDE LAFNEVLAKR SHKQLRATFQ AYQILIDKDI EEAIEAETSG DLQKAYLTLV RCARDQEGYF ADRLYKSMKG TGTDEETLIH IIVTRAEVDL QGIKAKFQEK YQKSLSDMVR SDTSGDFQKL LVALLH | |
Greater than 90% as determined by SDS-PAGE. |
403935 | |
100 μg | |
Not determined. | |
Full Length | |
Dog | |
His | |
Tris-based buffer, 50% glycerol |