missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Dengue Virus Dengue Epitope 10 Protein
A cDNA sequence encoding the Dengue Epitope 10 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11639-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
Dengue Epitope 10 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Phosphate buffered saline, pH 7.4 and 0.02% sodium nitrate. |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Dengue Virus | |
Untagged | |
Protein is >95% pure as determined by 12% PAGE (coomassie staining). |